VWR® Weighing Paper
Supplier: VWR International
12578-121 12578-165 12578-201
12578-121CS 3282.62 CAD
12578-121 12578-165 12578-201
VWR® Weighing Paper
Weighing Papers
Weighing paper is designed for analytical weighing with minimal influence on analytical results for weighing and transporting sample.
  • Clean and dust-free
  • Lightweight
  • Nitrogen-free
  • Smooth surface
A smooth surface is ideal for transferring weighed material. Clean, dust-free sheets. Paper is lightweight, yet strong enough to support analytical size samples.
Packaging: All sizes of weighing papers are packaged 500 sheets per box.