Sanitary Heavy-Duty Floor Squeegees, Perfex
Supplier: Perfex
9518 9524 9530
22940-032EA 91.73 CAD
22940-032 22940-034 22940-036
Sanitary Heavy-Duty Floor Squeegees, Perfex
Mops
These 100% polymer construction floor squeegees are nonsparking and chemical-resistant
Twin-mounted PVC blades act as a splash guard and are easily changed with the self-locking connection system. Lightweight, durable blade and handle will not absorb bacteria, liquids, or odor. Unaffected by water, petroleum products, detergents, sanitizers, and solvents. Squeegees require polymer handle (sold separately). Polymer handle will not splinter, fatigue, or conduct electricity.
Order Now
SPECIFICATIONS
Environmentally Preferable