Quick-Dry Foam Squeegees, Perfex
Supplier: Perfex
9724 9718
11311-118CS 986.76 CAD
11311-118 11311-120
Quick-Dry Foam Squeegees, Perfex
Mops
Twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces
Note: Not recommended for abrasive surfaces.
Feature a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, nonmarring, and easy to clean with warm water. Resistant to water, oil, grease, detergents, sanitizers, and solvents.
Ordering information: Polymer adjustable handle sold separately.