PIG® Absorbent Packaging Bag
Supplier: New Pig
PKG017 PKG018 PKG019 PKG020 PKG021
77463-550BX 114.12 CAD
77463-550 77463-552 77463-554 77463-556 77463-558
PIG® Absorbent Packaging Bag
Bags Storage Bags
Secure your shipment from escaping liquid by sealing your container in this packaging bag with absorbent sheet.
  • Sealable plastic bag with absorbent sheet traps and soaks up liquid escaping from container
  • Soft-but-heavyweight sheet adds light cushioning and scratch protection
  • Built-in mat flap lets you fold mat over the top for added barrier from seal point; tab also makes it easy to find the opening
  • Absorbs up to 1.41 ounces per bagged absorbent
  • Absorbent-bag combination works on most free liquids, including solvents, oils and water-based liquids
  • Durable
More Product Information