KIMAX® Separatory Funnels, Cylindrical, Graduated, Double Scale, PTFE Stopcock, [ST] Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
29040F-1000 29040F-125 29040F-250 29040F-500
89002-074CS 1495.11 CAD
89002-074 89002-076 89002-078 89002-080
KIMAX® Separatory Funnels, Cylindrical, Graduated, Double Scale, PTFE Stopcock, [ST] Stopper, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Funnel is supplied with a [ST] ground glass stopper and a PTFE stopcock
Graduated with a double white ceramic enamel scale. Scale graduations on the right side are ascending and on the left side are descending. Calibrated to contain.
Certifications: Designed from ASTM Specification E1096, Type 1C requirements.
Order Now
SPECIFICATIONS
Stem length 60 mm
Environmentally Preferable