Vectra® Alpha® 10 and Alpha® 10 Cleanroom Wipers, Texwipe®
Supplier: Texwipe
TX1010 TX1012
TWTX1010CS 1710.57 CAD
TWTX1010 TWTX1012
Vectra® Alpha® 10 and Alpha® 10 Cleanroom Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These 100% continuous-filament polyester wipers are constructed in a double-knit, no-run, interlock pattern that is excellent for cleaning process equipment.
  • All four cut edges of the wiper are thermally sealed
  • Border seal provides a protective edge around the body of each wiper for minimum particle generation
Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments. To reduce particulation, wiper TWTX1010 is Vectra processed and wiper TWTX1012 is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Ordering information: Wipers are packed in SolventSafe Bag-Within-A-Bag® packaging.