Vectra® AlphaSorb® 10 and AlphaSorb® 10 Wipers, Texwipe®
Supplier: Texwipe
TX1052 TX1050
TW-TX1052CS 1790.53 CAD
TW-TX1052 TWTX1050
Vectra® AlphaSorb® 10 and AlphaSorb® 10 Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Two-ply double-knit 100% continuous-filament polyester wipers offer high sorbency performance.
  • All four edges are thermally sealed, forming a border to prevent particle and fiber release
Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments. To reduce particulation, wiper TWTX1050 is Vectra processed and wiper TW-TX1052 is laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Ordering information: SolventSafe Bag-Within-A-Bag® packaging.