Multi-Purpose Brushes, Perfex
Supplier: Perfex
3040B 3040G 3040R 3040W 3040Y
22940-094EA 170.4 CAD
22940-094 22940-284 22940-096 22940-100 22940-286
Multi-Purpose Brushes, Perfex
Brushes
These multi-purpose brushes contain moderately stiff polyester fiber.
  • Surround-set and flared fiber maintains surface contact at any angle and pprevents brush block from damaging surfaces
  • Durable fiber maintains shape and is visually coded to isolate areas of use
  • Impact-resistant polymer brush block withstands soaking and will not rot, decay, chip or crack
  • Non-conductive with superb abrasion and chemical resistance
  • Unaffected by water, grease, petroleum products, detergents, sanitizer, and solvents
Excellent for scrubbing irregular surfaces, as wellMore Product Information