KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
29048F-30 29048-1000 29048-125 29048-250 29048-500 29048-60 29048F-1000 29048F-125 29048F-2000 29048F-250 29048F-500 29048F-60 29048T-125 29048T-2000 29048T-250 29048T-500 29049F-1000 29049F-125 29049F-2000 29049F-250 29049F-500
30352-142CS 989.65 CAD
30352-142 89002-088 89002-090 89002-092 89002-094 89002-096 89002-098 89002-100 89002-102 89002-104 89002-106 89002-108 89002-110 89002-112 89002-114 89002-116 89002-118 89002-120 89002-122 89002-124 89002-126 89002-128
KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Funnels are designed with a large lower stem diameter to allow more complete separation.
  • Pear-shaped
  • Autoclavable
Constructed with heavy walls, a fire-polished and chip-resistant stem, and heavily beaded rim.
Certifications: Designed from ASTM Specification E1096, Type 4 requirements. Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with a Standard Taper ground glass stopper and an autoclavable PTFE stopcock plug.