KONTES® Separatory Funnels, Squibb, with Glass Plug Stopcock, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
636000-0030 636000-1000 636000-0060 636000-0125 636000-0250 636000-0500 636000-2000 636010-0060 636010-0250 636010-2000
KT636000-0030EA 253.45 CAD
KT636000-0030 KT636000-1000 KT636000-0060 KT636000-0125 KT636000-0250 KT636000-0500 KT636000-2000 KT636010-0060 KT636010-0250 KT636010-2000
KONTES® Separatory Funnels, Squibb, with Glass Plug Stopcock, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Squibb-type funnels feature a [ST] joint and a [ST] glass plug stopcock
Plain stem. Available with or without a [ST] stopper.