Polynit Wipers, Contec®
Supplier: Contec
PN-99 PN-44 PN-44B PN-99 B PN-1212 PN-1818
68300-558CS 1100.04 CAD
68300-558 68310-166 68310-168 68310-172 68310-186 77507-160
Polynit Wipers, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polynit Wipes are made of 100% knitted textured polyester with knife cut edges for a very soft wipe. The soft edge is ideal for use on surfaces which are vulnerable to scratching.
  • Recommended for ISO Class 5 to 6 environments
  • Laundered knitted 100% polyester fabric
  • Soft edge is ideal for use on surfaces which are vulnerable to scratching
  • 100% knitted polyester is compatible with many solvents
  • Durable fabric is abrasion and chemical resistant
Polyester wipes are chemically resistant and exceptionally low in particles and fibers. An interlock knit creates a durable fabric, the addition of a periodic additional stitch of a no-run interlock knit prevents the fabric unraveling creating an even more durable fabric with strong edges.
The wipes are laundered and packaged in an ISO Class 4 cleanroom. They have good sorbency with solvents and are abrasion and chemical resistant. A validated sterile version is available.