Super Polx® 1200 Wipers, Berkshire
Supplier: Berkshire
S12000404.10 S1200.0909.8 S1200.0909B.8
21914-401CS 1601.19 CAD
21914-401 21914-445 21914-467 21914-474
Super Polx® 1200 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durable, 100% knitted polyester wipers for general cleaning purposes, spill pickup, and equipment cleaning.
  • Low particles, fibers, ions, and extractable
  • High abrasion resistance
  • Chemically compatible with IPA, acetone, and other solvents
  • Soft hand and edge for sensitive surfaces
Continuous-filament, no-run construction offers resistance to chemicals and abrasion. Low extractables, with extremely low particle and fiber generation. For use in ISO Class 4 (FED-STD-209E Class 10/M2.5) environments. Manufactured and tested with strict adherence to qualityMore Product Information