Polx® 1200 Wipers, Berkshire
Supplier: Berkshire
P12000909B.1 P12000404.18 P1200.0909.8 P12001212.14
10810-650CS 898.4 CAD
10810-650 21914-300 21914-332 21914-354
Polx® 1200 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Well-suited to optical applications and use on other sensitive surfaces, these all-purpose wipers are an economical midpoint between nonwoven materials and laundered synthetic wipers.
  • Low particles, fibers, ions, and extractable
  • High abrasion resistance
  • Chemically compatible with IPA, acetone, and other solvents
  • Soft hand and edge for sensitive surfaces
Wipers are made of durable, yet nonabrasive 100% double-knit polyester. They are low in particle generation and extractables, making them useful in applications that would abrade or shred a nonwoven material. Recommended for ISO Class 5 (FED-STD-209E Class 100/3.5) environments.