Glucose Tolerance Drinks, Azer Scientific
Supplier: Azer Scientific
10-O-050-CA 10-O-075-CA
CA11006-157CS 238.24 CAD
CA11006-157 CA11006-153 CA11006-155
Glucose Tolerance Drinks, Azer Scientific
Beverages
FDA-approved, non-carbonated beverages are designed for use in conjunction with glucose tolerance tests.
  • Shatterproof Plastic Bottles
  • Graduated Labels on Bottles
  • Non-Carbonated Beverages
  • Citric Acid Free
  • Dairy and Gluten-Free
All beverages are supplied in shatterproof plastic bottles.
Beverages are gluten- and dairy-free, certified Kosher, and are manufactured according to WHO and ADA standards. Simply Pure unflavored beverage is also citric acid free. The integrity of each batch is certified by an FDA recognized independent testing laboratory. All bottles are marked with an expiration date determined via accelerated and real time stability studies.
A graduated label on each bottle indicates the amount dispensed.
Certifications: Beverages are gluten and dairy free, certified Kosher, and are manufactured according to WHO and ADA standards. Simply Pure unflavored beverage is also citric acid free. The integrity of each batch is certified by a FDA-recognized independent testing laboratory. All bottles are marked with an expiration date determined via accelerated and real-time stability studies.
Ordering information: Beverages are packaged in four shrink-wrapped six-packs per case. Beverages are packaged in four shrink-wrapped six-packs per case.