SP Bel-Art Sterileware® Economy Sterile Sample Spoons, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36740-0001 F36740-0002 F36740-0003 F36740-0004 F36740-0005 F36740-0310
74950-181CS 254.89 CAD
74950-181 74950-183 74950-185 74950-187 74950-189 89140-568 89140-570 89140-572 89140-574 89140-576
SP Bel-Art Sterileware® Economy Sterile Sample Spoons, Bel-Art Products, a part of SP
Spoons
These economical measuring devices are ideal for retrieving small, specific volume samples of powders or liquids.
  • Economical
  • Stiff and durable for sample packed powders
  • Polystyrene plastic construction
  • Flat-bottomed shape
Their unique flat-bottom shape allows them to be used as weighing boats for additional sampling accuracy.
Use a separate spoon for each material sampled to avoid cross contamination. All sizes are made from white, FDA-grade high-impact polystyrene plastic for durability and stiffness when sampling packed powders. Supplied sterileMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable