AlphaSorb® HC Wipers
Supplier: Texwipe
TX2412 TX2418 TX2409
TWTX2412CS 1449.74 CAD
TWTX2412 TWTX2418 TW-TX2409
AlphaSorb® HC Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
AlphaSorb® HC (high capacity) two-ply, knitted, 100% polyester wipers are engineered to combine high sorbency with low particle and extractable levels.
  • Autoclavable
The large surface area is especially suited for use in the pharmaceutical and biotechnology industries where large, highly sorptive wipers are required for applying and removing cleaning, sterilizing, and disinfecting solutions.
Certifications: Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
Ordering information: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom and packed in SolventSafe Bag-Within-A-Bag® packaging.