Wide-Mouth Packers, Amber, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216939 W216940 W216941 W216942 W216943 W216944 W216945 W216946 W216947 W216948 W216949 W216950 W216951 W216952 W216953 W216954 W216955 W216956 W216957 W216958 W216959 W216960 W216961 W216962 W216963 W216964 W216965 W217962 W217963 W217964 W217965 W217960 W217961
14235-890CS 46.04 CAD
14235-890 14235-892 14235-894 14235-896 14235-898 14235-900 14235-902 14235-904 14235-906 14235-908 14235-910 14235-912 14235-914 14235-916 14235-918 14235-920 14235-922 14235-924 14235-926 14235-928 14235-930 14235-932 14235-934 14235-936 14235-938 14235-940 14235-942 10847-486 10847-488 10847-490 10847-552 10847-686 10847-688
Wide-Mouth Packers, Amber, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton wide-mouth packers are manufactured from amber soda lime glass that conforms to USP Type III requirements.
Amber glass provides protection for light sensitive products. The wide-mouth design is ideal for use with dry powders. A partitioned carton protects bottles from scratches during shipping. Each case also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Available with caps attached or in bulk packs without caps. Replacement screw caps can be purchased separately.