Standard Wide Mouth Bottles, Clear, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216924 W216925 W216926 W216929 W216930 W216931 W216933 W216934 W216935 W216936 W216937
14235-860CS 70.39 CAD
14235-860 14235-862 14235-864 14235-866 14235-868 14235-870 14235-872 14235-874 14235-876 14235-878 14235-880 14235-882 14235-884 14235-888 14235-886
Standard Wide Mouth Bottles, Clear, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton standard wide mouth clear bottles are manufactured from clear soda-lime glass that conforms to USP Type III requirements.
These wide mouth Boston round bottles are offered with a choice of PTFE faced foam polyethylene liner with polypropylene cap, or poly-vinyl liner with polypropylene cap.
Bottles are supplied with caps attached or without caps.
The bottles are ideal for general-purpose, small to large volume storage. Caps are available with the choice of two different liners. Bottles are packaged in convenience packs with or without caps attached. Caps may also be purchased separately.