Cleanroom Pens and Markers
Supplier: Micronova
PEN-01IR PEN-02IR PEN-20IR PEN-25IR PEN-22IR PEN-11 PEN-11IR PEN-12 PEN-12IR PEN-52IR PEN-51IR PEN-20IRBB PEN-25IR PEN-61IR
68400-568CS 471.12 CAD
68400-568 68400-570 68400-572 89032-008 89107-766 89236-620 76203-464 76214-024 76214-026 76214-028 76214-296 76292-770 76443-120 76214-299 89107-767 76471-004 76292-771
Cleanroom Pens and Markers
Pens Cleanroom Pens
Micronova cleanroom pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom Irradiated Sharpie® and Smear-Proof markers write permanently on most surfaces with quick drying ink that is fade- and water-resistant to further extend multiple project uses. Marks will not smear on reusable glass, metal, or porcelain lab-ware, but can be scrubbed off.
  • Indelible, low sodium ink pens
  • White, high-density polypropylene barrel
  • Holds up to IPA, bleach, and phenol wipe downs
  • Available capped or retractable
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable