Quiltec® I Wipers
Supplier: Contec
LWQS0001 LWQS0002 QT1-99 QT1-44B QT1-99B QT1-1212 QT1-1212B/100 LWQT0019
68311-024CS 1471.02 CAD
68311-024 68311-025 68300-560 68310-215 68310-218 68310-222 89074-012 75996-814
Quiltec® I Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Quiltec® I Wipes offer the highest sorptive capacity of any sealed edge wipe available. They are manufactured using two layers of knitted polyester fabric that are laminated together to form a stronger, more adsorbent wipe. Additional fluid is captured within the quilted pockets between the fabric layers, increasing the sorbent capacity. Quiltec® I have more than twice the adsorbent capacity of a single-layer wipe.
  • Extremely low in both particles and fibers, reducing possibility of contamination
  • Strong fabric is abrasion and chemical resistant
  • Excellent sorbency with solvents
These wipes have laser-cut, heat-sealed edges which reduce the number of releasable particles and fibers. They are laundered and packed in a class 4 cleanroom. Sterile Quiltec® wipes are sterilized via gamma irradiation, and a certificate of sterility is provided with each shipment.
Certifications: Recommended for ISO Class 3-5 environments.