Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
R1489000-4A R1489000-1A R1490000-4A R1490000-1A R1490000-500A R1492000-4A R1492000-1A R1493000-4A R1493000-20F R1495000-4A R1495000-1A R1495000-20F R1498000-4A R1498000-1A R1498000-500A R1500000-4A R1500000-10F R1500000-1A R1500000-20F R1505000-4A R1505000-1A R1510000-4A R1510000-1A R1540000-4A R1540000-1A R1540000-500A R1550000-4A R1550000-500A R1550000-10F R1550000-1A R1550000-20F R1489000-500A R1492000-500A R1493000-500A R1493000-1A R1495000-500A R1500000-500A R1505000-500A R1510000-500A R1550000-4F 1485-1 1485-16 1485-32 R1492000-20PK R1493000-20PK R1489000-20F R1492000-10F R1493000-10F R1495000-10F R1495000-20PK R1498000-20F R1505000-10F R1505000-20F R1510000-10F R1540000-10F R1540000-20F R1565000-1A R1565000-20F R1565000-4A R1565000-500A
RC1489-1EA 385.52 CAD
RC1489-1 RC1489-32 RC1490-1 RC1490-32 RC149016 RC1492-1 RC1492-32 RC1493-1 RC1493-5 RC1495-1 RC1495-32 RC1495-5 RC1498-1 RC1498-32 RC149816 RC15001 RC150025 RC150032 RC15005 RC1505-1 RC1505-32 RC1510-1 RC1510-32 RC1540-1 RC1540-32 RC154016 RC15501 RC155016 RC155025 RC155032 RC15505 RC1489-16 RC149216 RC1493-16 RC1493-32 RC1495-16 RC150016 RC1505-16 RC1510-16 RC1550-1CT RC1485-1 RC1485-16 RC1485-32 RCR149200020PK RCR149300020PK RCR1489000-20F RCR1492000-10F RCR1493000-10F RCR1495000-10F RCR149500020PK RCR1498000-20F RCR1505000-10F RCR1505000-20F RCR1510000-10F RCR1540000-10F RCR1540000-20F RCR1565000-1A RCR1565000-20F RCR1565000-4A RCR1565000500A
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers pH Calibration Buffers
Colorless pH buffer solutions are certified to ±0.01 pH at 25°C
  • Range of packaging is available
Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.
Certifications: Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.