Microplate Lids and Seals
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
202497-100 200856-100
CA77463-680PK 358.13 CAD
CA77463-680 CA77463-660
Microplate Lids and Seals
Lids Microplate Lids
Universal lids and cap mats for square microplates.
  • Universal-sized polystyrene lids fit most all microplates
  • Black color is effective for light-sensitive reagents
  • Robot-friendly design
  • Cap mats specifically designed for optimal fit for Agilent square-well microplates
  • Mat material has low permeability to alcohols
Agilent microplate accessories include microplate lids and cap mats. Agilent supplies universal lids for microplates, available in clear or black polystyrene resin. Custom cap mats are available for sealing Agilent 24, 48, and 96-square well microplates. These custom cap mats are molded in Santoprene, a chemical-resistant thermoplastic elastomer.