ValuSeal-HA® Sealed-Edge Cleanroom Wipers, Berkshire
Supplier: Berkshire
VSHA.0909.8 VSHA.1212B.14
80094-518CS 1523.14 CAD
80094-518 80094-520 80094-522 77531-240
ValuSeal-HA® Sealed-Edge Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Highly absorbent, 100% knitted polyester wipers have ultrasonic heat-sealed edges.
  • Lightweight
  • Low particles, fibers, ions, and extractable
  • High absorbency material reduces actual cost in use compared to heavier weight products
Wipers work well with IPA, acetone, and other solvents. Steam autoclavable for aseptic environments.
Certifications: Recommended for ISO Class 3 (FED-STD-209E Class 1/M1.5) cleanroom environments, including semiconductor and biomedical cleanroom applications.
Ordering information: Wipers are laundered and packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.