VWR® Sample Bags for the Seward Stomacher® Blender
Supplier: VWR International
CA11216-898 CA89085-562 CA11216-890 CA11216-892 CA11216-900 CA11216-902 CA89003-820 CA89085-564 11216-892 89085-564 10048-880 10048-882 10048-884 10048-886 10048-888 10048-890 SCTO7012A-VW EFT-6090A-VW SCT-7012A-VW EFT-7012A-VW SCT-1520A-VW EFT-1015A-VW CA89085-567 76200-884
CA11216-898CS 529.76 CAD
CA11216-898 CA89085-562 CA11216-890 CA11216-892 CA11216-900 CA11216-902 CA89003-820 CA89085-564 11216-892 89085-564 10048-880 10048-882 10048-884 10048-886 10048-888 10048-890 CA10048-880 CA10048-882 CA10048-884 CA10048-886 CA10048-888 CA10048-890 CA89085-567 76200-884
VWR® Sample Bags for the Seward Stomacher® Blender
Bags Blender Bags
Sample bags are designed to withstand the paddle action of Stomacher® lab blenders.
  • Pure or hybrid polyethylene construction
  • Highly transparent
  • Suitable for general-purpose blending or analysis testing
  • Some bags feature write-on strip
  • Malleable yet durable
FILTRA-BAG® bags are made of a mix of polyethylene and nylon.
SECURE-T® bags are made of heat extruded polyethylene tubing for internal sterility and to eliminate the need for side seals. Bags are malleable yet durable, with superior wall strength. May be used in general-purpose blending or analysis testing.
The LABRACK® storage rack, an ideal tool for storing samples, is designed to store up to ten 400 series blender bags, closed with a clip, in their upright position.
All standard and closure bags are made of highly transparent polypropylene and have wide-sealed bottoms to prevent leaks. Bags with 4 mm wall thickness are for analyses of firmer samples. Standard bags are made with heat-extruded virgin polyethylene tubing and feature a tear-off sterile barrier top to ensure sterility up to the time of use. Closure bags permit collection, transportation, blending, and storage of samples all within the same bag. They are available with regular wire or extra-strength flat wire closures. Some bags feature a write-on strip for easier sample identification.