Straight Sided Round Bottles, Clear, Vacuum-Treated and Ionized, Qorpak
Supplier: Qorpak
GLC-01727 GLC-01597 GLC-01588 GLC-01613 GLC-01636 GLC-01674 GLC-01694 GLC-01723 248960 249705 249706 249707 249717 GLC-01646 GLC-09345 280505 280578 GLC-01699
11311-492CS 178.75 CAD
11311-492 16151-578 16151-584 36319-734 36319-739 36319-780 36319-781 36319-782 36319-783 36319-784 36319-785 76170-376 76170-378 76170-380 76170-382 76170-384 76487-330 76487-336 76487-338 76487-340 76487-342 76487-344 76487-346
Straight Sided Round Bottles, Clear, Vacuum-Treated and Ionized, Qorpak
Bottles Glass Bottles
The vacuum and ionized cleaning process removes contaminants such as loose dirt, carton lint, aerosols, and fine glass particles.
  • Bottles are made of glass
Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Then the air passes through an absorbent filter to remove gases. A vacuum then draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. The air inside the bottle will not interfere with your product in any way.