VWR® Crucibles, PTFE
Supplier: VWR International
89026-070 89026-072 89026-074 89026-076
89026-070EA 126.97 CAD
89026-070 89026-072 89026-074 89026-076
VWR® Crucibles, PTFE
Crucibles
Isostatically molded, pure FDA approved PTFE crucibles are totally chemically resistant and inert.
  • Chemically resistant
  • Inert
  • PTFE fabrication
For use in temperatures up to 270°C (518°F).