VWR® Nickel Crucibles and Covers
Supplier: VWR International
82027-556 82027-558 82027-560 82027-562 82027-564 82027-566
82027-556CS 4592.35 CAD
82027-556 82027-558 82027-560 82027-562 82027-564 82027-566
VWR® Nickel Crucibles and Covers
Crucibles
High form crucibles and covers are drawn from one-piece, pure sheet nickel.
They are highly resistant to dilute alkalies, and can withstand temperatures up to 800°C (1472°F).
Ordering information: Crucibles and covers are sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable