Composite Test Jar, Clear, Wide Mouth, Qorpak
Supplier: Qorpak
GLA-00852 GLC-01603 GLC-01600
16087-016CS 723.96 CAD
16087-016 16195-882 16195-884 76487-328
Composite Test Jar, Clear, Wide Mouth, Qorpak
Bottles Glass Bottles
Clear, flint glass composite test jars are wide-mouth, straight-sided jars that are ideal for sampling.
  • Provide maximum content visibility
Designed without shoulders for maximum storage capacity.