VWR® Total-Range Digital Thermometer
Supplier: VWR
VWR®
4015
23609-232EA 563.67 CAD
23609-232
VWR® Total-Range Digital Thermometer
Thermometers Digital Thermometers
Fast-reading, easy-to-use thermometer switches from °C to °F at the touch of a button.
  • Traceable®
  • Reads °C and °F
  • Large display with easily legible digits
  • Memory of minimum and maximum readings
Large display with 1.6 cm (5/8") high digits is legible from 3m (10') away
All Type-K probes may be used with liquids, air/gas, and semi-solids. Stainless steel probe has a 22.9 cm (9") stem with a 3mm (1/8") diameter and a 102 cm (401/8") cable. Beaded probe is 1.5mm (1/16") in diameter and features a 1.2m (4') fluoropolymer resin cable. The low-temperature stainless steel probe has a diameter of 4.3mm (3/16") and features a 30.5 cm (12") stem with a 12.7 cm (5") handle. Low-temperature probe is supplied with a 2.4m (8') expandable cable.
Thermometer recalls minimum and maximum readings from any time period. Alarm function may be programmed in 1° increments to sound when temperature rises above or falls below set points. Count-up timer alarm may be set from 23 hours, 59 minutes, to 1 minute. Unit is constructed of shock-resistant ABS plastic. Water-resistant PVC protective case stops water, fumes, dirt, and dust. Uses all Type-K thermocouple probes. Batteries power unit for one year of routine use.
Backlighting button permits reading in dark areas. Unit features a hold button that captures current reading. Microprocessor updates display twice per second.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate thermometer traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a PVC protective casing with flip-out stand, a fast-response Type-K beaded probe (61161-372), and three AAA alkaline batteries.