Diamond® Essentials™ Graduated Measuring Cylinders
Supplier: Globe Scientific
3701-1M 3701-250 3701-50 3701-500 3703-10 3703-1M 3703-25 3703-250 3703-2M 3703-50 3703-500 3705-100 3705-1M 3705-250 3705-2M 3705-500
77447-938EA 30.86 CAD
77447-938 77447-942 77447-946 77447-948 77447-950 77447-954 77447-956 77447-958 77447-960 77447-962 77447-964 77447-968 77447-970 77447-974 77447-976 77447-980
Diamond® Essentials™ Graduated Measuring Cylinders
Cylinders Measuring Cylinders
Graduated measuring cylinders offer the precise fluid measurements required in many laboratory operations. These cylinders are available in both Class A accuracy for precision work, and Class B accuracy for general laboratory work. Engineered with easy to read molded graduations, hexagonal bases, wide openings, and tapered pour spouts for simple and efficient use.
  • Available in translucent polypropylene (PP) or clear polymethylpentene (PMP)
  • Manufactured from virgin, USP class VI resins
  • Chemically resistant to most acids, bases, and many common solvents
  • Molded metric graduation scales per Class A or B accuracy standards
Working temperature range: 4 to 60 ºC. Calibrated to contain (TC) (In) at 20 ºC.
Certifications: Complies with FDA 21-CFR and European Pharmacopeia. Comply with DIN 12681 and ISO 6706 standards.