Shallow Form Watch Glasses, Walter Stern
Supplier: Walter Stern
110-100 110-101 110-102 110-103 110-104 110-105 110-106 110-107 110-108 110-110 110-112 110-114
66112-027PK 175.93 CAD
66112-027 66112-049 66112-060 66112-082 66112-107 66112-129 66112-140 66112-162 66112-184 66112-209 66112-220 66112-242
Shallow Form Watch Glasses, Walter Stern
Watch Glasses
Concave, clear, annealed glass covers have fire-polished edges
Used to return condensation to beakers. Manufactured from soda lime glass.