Blotter Paper, IW Tremont
Supplier: I W TREMONT CO CA
IWT-BLOT88
CAIWT-BLOT88CS 1009.49 CAD
CAIWT-BLOT88
Blotter Paper, IW Tremont
Blotting Papers
Blotter paper with smooth surface. Made from bright white cellulose filter paper. Material is manufactured from carefully selected, fully bleached and chemically free pure cellulose fiber without additives or binder.
Meets standards specified in TAPPI T 205 sp-06.