PYREX® Gooch Crucibles, Low Form, with Fritted Disc, Corning
Supplier: Corning
32960-30F 32960-30C 32960-30M
23872-063CS 1318.75 CAD
23872-063 89091-332 89091-334
PYREX® Gooch Crucibles, Low Form, with Fritted Disc, Corning
Crucibles
Particularly adapted to analytical work in cases where precipitates are dried to constant weight at 110 °C (230 °F).
  • Made of PYREX® borosilicate glass
Also suitable for high temperatures, in which case it is advisable to heat in an electric furnace. In order to avoid strain, crucible should not be removed from furnace until the temperature has dropped to 250 °C (482 °F). Limit of use, 500 °C (932 °F).