CleanMax® Cleanroom Frocks
Supplier: Lakeland
CTL191CMB-SM CTL191CM-SM CTL191CS-SM
77449-030CS 329.68 CAD
77449-030 77449-032 77449-034
CleanMax® Cleanroom Frocks
Disposable Garments
Cleanroom garments are constructed and packaged to offer premium protection for your cleanroom and controlled environments.
  • Available in sterile and non-sterile configurations
  • Frock with zipper closure and collar
  • Bound seam
CTL191CM (clean manufactured) and CTL191CS (clean manufactured and sterile) cleanroom frock is ideal for ISO Class 4 to 6 cleanrooms.
Delivery information: These products are offered in a case of 30 with sizes small to 5XL available.