Ethyl acetate, anhydrous 99,8, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid ethyl ester

Septum-Sealed Bottle

EX0237-6 EX0237-7
CAEX0237-6CS 823 CAD
CAEX0237-6 CAEX0237-7
Ethyl acetate, anhydrous 99,8, DriSolv®, Supelco®
Ethyl acetate

Flammable.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Residue after evaporation 1ppm max
Water (H2O) 50ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR