Diethyl ether, anhydrous ≥99% stabilized, DriSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
Packed in septum-sealed bottle.
Stabilized with 1ppm BHT. Packed under nitrogen. Flammable.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.
Formula:
(CH₃CH₂)₂O MW: 74.12 g/mol Boiling Pt: 35 °C (1013 hPa) Melting Pt: –123 °C Density: 0.7135 g/cm³ (20 °C) Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
Specification Test Results
Expiration date | Exp 24 months from mfg date |
Assay (GC) | 99.8% min |
Appearance | Clear liquid, free from particulates |
Color (APHA) | 10 max |
Peroxide (as H2O2) | 1ppm max |
Residue after evaporation | 3ppm max |
Water (H2O) | 50ppm max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...