Tetrahydrofuran, anhydrous ≥99.8%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Packed in septum-sealed bottle.

TX0284-7 TX0284-NP20
CATX0284-7CS 726 CAD
CATX0284-7 CATX0284-NP20
Tetrahydrofuran, anhydrous ≥99.8%, DriSolv®, Supelco®
Tetrahydrofuran

Clear and free from particulates. Stabilized with 25ppm BHT. Suitable for use in organic synthesis and other anhydrous solvent applications. Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid, free from particulates
Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Stabilizer (BHT) ~25ppm min
Water (H2O) at time of manufacture 20ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR