KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17121H-25 17121H-50
89001-648CS 270.47 CAD
89001-648 89001-650
KIMAX® Burets with Threaded Closure Fit, Straight Bore PTFE Stopcock, Kimble Chase
Burettes
Burets are composed of four replaceable components and incorporate a threaded closure that eliminates accidental dislodging of the stopcock assembly during use
Note: Assembly should be wetted before installation.
The graduated tube has a reinforced top with a lower end accurately tooled to provide precise I.D. fit with a PTFE stopcock assembly. A ground tip fits into the lower end of the stopcock assembly.
Ordering information: Burets include a KIM-KAP® dust cap to secure to the threaded buret body with a positive closure.