Pyridine, anhydrous ≥99.9%, DriSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Packed in septum-sealed bottle.
Suitable for use in organic synthesis and other anhydrous solvent applications. Clear liquid is filtered through a 0.2µm filter, making it free from particulates. Flammable and toxic.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septa (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.
Formula:
C₅H₅N Boiling Pt: 115 °C (1013 hPa) Melting Pt: –44 °C Density: 0.983 g/cm³ (20 °C) Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 13,08060 |
|
Specification Test Results
Assay (GC) | 99.8% min |
Appearance | Clear liquid, free from particulates |
Filtered through 0.2 µm filter | To pass test |
Residue after evaporation | 5ppm max |
Water (H2O) | 50ppm max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...