Methanol, anhydrous ≥99%, DriSolv®, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Packed in septum-sealed bottle.
Clear and free from particulates. Suitable for use in organic synthesis and other anhydrous solvent applications. Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.
Formula:
CH₃OH Boiling Pt: 64.6 °C (1013 hPa) Melting Pt: –98 °C Density: 0.7918 g/cm³ (20 °C) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Specification Test Results
Assay (GC) | 99.8% min |
Appearance | Clear liquid, free from particulates |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Residue after evaporation | 1ppm max |
Water (H2O) | 50ppm max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...