Dimethyl sulfoxide, anhydrous ≥99.8%, DriSolv®, Supelco®
Supplier: MilliporeSigma
Synonyms:
Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane
Packed in septum-sealed bottle.
Clear and free from particulates. Suitable for use in organic synthesis and other anhydrous solvent applications.
Accessories information: Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2), filled with t.h.e.* desiccant indicating silica gel, removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.
Formula:
(CH₃)₂SO MW: 78.14 g/mol Boiling Pt: 189 °C (1013 hPa) Melting Pt: 18.5 °C Density: 1.101 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
Specification Test Results
Appearance | Clear liquid, free from particulates |
Assay (GC) | 99.8% min |
Color (APHA) | 10 max |
Residue after evaporation | 5ppm max |
Filtered through 0.2 µm filter | To pass test |
Water (H2O) at time of manufacture | 50ppm max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...