N,N-Dimethylformamide, anhydrous ≥99.8%, DriSolv®, Supelco®

Supplier: MilliporeSigma

Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

Packed in septum-sealed bottle.

DX1727-6 DX1727-7 DX1727-NP20 DX1727-1 DX1727-6 DX1727-NP20 DX1727-7 DX1727-1
CADX1727-1CS 6198.79 CAD
CADX1727-1 EM-DX1727-NP20 EM-DX1727-1 EM-DX1727-7 CADX1727-NP20 EM-DX1727-6 CADX1727-7 CADX1727-6
N,N-Dimethylformamide, anhydrous ≥99.8%, DriSolv®, Supelco®
N,N-Dimethylformamide

Clear liquid is free from particulates. Suitable for use in organic synthesis and other anhydrous solvent applications.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapter, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
MDL Number: MFCD00003284
CAS Number: 68-12-2
UN: 2265
ADR: 3,III
Merck Index: 13,03269
https://ca.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://ca.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg

Order Now

Specification Test Results

Appearance Clear liquid, free from particulates
Assay (GC) 99.8% min
Color (APHA) 10 max
Residue after evaporation 5ppm max
Filtered through 0.2 µm filter To pass test
Water (H2O) at time of manufacture 50ppm max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR