Dichloromethane, anhydrous ≥99.8% stabilised, DriSolv®, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Methylene chloride
Stabilization:
Stabiliser
Packed in septum-sealed bottle.
Clear and free from particulates. Suitable for use in organic synthesis and other anhydrous solvent applications.
Elbow adapter with septum cap (EM-692004-2) allows for the removal of solvent under nitrogen atmosphere. Replacement septum (EM-692004S-2), for use with elbow adapters, minimizes air and moisture intrusion once the primary septum is punctured. Drying tube (EM-692010-2) filled with t.h.e.* desiccant indicating silica gel removes solvent under a dry-air atmosphere. Large plug (EM-692012-2) is a replacement plug for bottle top.
Formula:
CH₂Cl₂ MW: 84.93 g/mol Boiling Pt: 39.8 Melting Pt: -95 Density: 1.322 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1 III Merck Index: 12,06140 |
|
Specification Test Results
Assay (GC) | 99.8% min |
Appearance | Clear liquid, free from particulates |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Residue after evaporation | 1ppm max |
Water (H2O) | 50ppm max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...