Low-Particle Bottles and Jugs, Thermo Scientific
Supplier: Thermo Scientific
114-125CT/LP 114-060A/LP 110-80A/LP 111-04A/LP 113-500A/LP 114-125A/LP 114-250A/LP 114-250C/LP
89026-620CS 384.82 CAD
89026-620 89026-624 EP110-80A-LP EP111-04A-LP EP113-500ALP EP114-125A-LP EP114-250A-LP EP114-250C-LP
Low-Particle Bottles and Jugs, Thermo Scientific
Bottles Glass Bottles
Glass containers are certified to contain fewer than 5 particles per ml larger than 0.5 µm.
  • Class 100/10 glass
  • Certificate of Analysis is included with each shipment
  • Available in clear or amber
  • Individual bottles double bagged to maintain integrity until point of use
  • 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
  • Recommended for low-particle applications
Ideal for preparation and storage of standards, off-line D.I. water and chemical sampling for particle analysis, and storage and packaging of chemicals, reagents, and powders. Amber glass protects light-sensitive chemicals. Containers are assembled with low-shedding, polypropylene caps with chemically-inert fluoropolymer-resin faced liners that do not contain adhesives. Exceeds USP specifications for particle content of containers.
Ordering information: All containers are double-bagged in a ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom. Supplied with Certificate of Analysis.