EP Scientific Low-Carbon Total Organic Carbon Vials, Tubes and Bottles, Thermo Scientific
Supplier: Thermo Scientific
40A-TOC/DB/LL 40C-TOC 40C-TOC/DB 40C-TOC/DB/LL 40C-TOC/LL CT18-TOC/LL S114-125A/TOC S114-250A/TOC S114-250C/TOC SCT-18100/TOC 1000A/TOC 3115-OTWS-2
EP40A-TOCDBLLCS 2304.08 CAD
EP40A-TOCDBLL EP40C-TOC EP40C-TOC-DB EP40C-TOC-DBLL EP40C-TOC-LL EPCT-18-TOC-LL EPS114-125ATOC EPS114-250ATOC EPS114250CTOC EPSCT18100TOC EP1000A-TOC 10142-542
EP Scientific Low-Carbon Total Organic Carbon Vials, Tubes and Bottles, Thermo Scientific
Bottles Glass Bottles
Containers are certified to contain low levels of carbon and are for use in Total Organic Carbon (TOC) testing and sampling. Ideal for cleaning validation, off-line sampling and grab sampling of high purity water, and for preparation and storage of standards.
  • Available in several sizes and styles of containers, including the popular 40 ml autosampler vials
  • Cleaned, certified and ready to use
All containers have open-top caps, unless otherwise specified, and fluoropolymer resin-lined septa.
Manufactured, tested and packaged by exacting standards,More Product Information