PYREX® Micro Burets, Class A, Funnel Top, Precision Bore, Straight [ST] Stopcock, Corning
Supplier: Corning
2128-10 2128-5
89091-092CS 2393.04 CAD
89091-092 89091-478
PYREX® Micro Burets, Class A, Funnel Top, Precision Bore, Straight [ST] Stopcock, Corning
Burettes
Micro burets, Class A, PYREX® borosilicate glass.
  • Precision bore tubing
  • Funnel top
  • Tapered tips for smooth flow
  • Stopcocks turn freely yet give positive control
Designed for precise analytical methods in microchemistry. 
Certifications: The capacity tolerance on these burets is established by ASTM E1189 and are calibrated in accordance with ASTM E542.