PYREXPLUS® Safety-Coated Reagent Bottles, Narrow Mouth, Corning
Supplier: Corning
61500-125 61500-250 61500-1L 61500-2L 61500-500
16266-221CS 1766 CAD
16266-221 16266-224 89091-666 16266-232 89091-668
PYREXPLUS® Safety-Coated Reagent Bottles, Narrow Mouth, Corning
Bottles Glass Bottles
PYREX® borosilicate glass, coated.
  • Conventional bottle shape
  • Autoclavable at 121 °C and resistant to thermal shock
  • Used for storage of solutions, ideal for media preparation
Designed with protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Neck is tooled to increase mechanical strength and for uniform stopper fit.
Not recommended for dry heat sterilization as plastic coating will melt. Can be written on with a standard lab marker, permanent or water-soluble.