KIMAX® Burets, Class B, Straight Bore Stopcock, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17026F-10 17026F-100 17026F-25 17026F-50
89001-606CS 78.88 CAD
89001-606 89001-608 89001-610 89001-612 89001-614
KIMAX® Burets, Class B, Straight Bore Stopcock, Kimble Chase
Burettes
Burets feature a 2mm stopcock plug and a KIM-KAP® dust cap
Scale is printed in durable black ceramic enamel. Designed from ASTM Specification E287, Class B requirements.