EP Scientific Depyrogenated Vials, Sterile, Thermo Scientific
Supplier: Thermo Scientific
ST10-20 ST100-20 ST2-13 ST20-20 ST30-20 ST5-20 ST50-20 ST100-20A
EPST10-20CS 594.46 CAD
EPST10-20 EPST100-20 EPST2-13 EPST20-20 EPST30-20 EPST5-13 EPST5-20 EPST50-20 10510-820
EP Scientific Depyrogenated Vials, Sterile, Thermo Scientific
Bottles Glass Bottles
Type I borosilicate vials are suitable for a number of uses where aseptic protocol is required.
  • Vials are assembled with butyl stoppers and aluminum seals
Certifications: Sterilization is performed in a FDA licensed facility.
Ordering information: Supplied with Certificate of Sterility and Pyrogen Test.