Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
3640-0010 3640-0025 3640-0050
17589-001CS 1165.89 CAD
17589-001 17589-056 17589-103
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Burettes
Excellent for industry and field
Meets ASTM E287 for accuracy.
Automatic self-zeroing burets are individually calibrated for superior accuracy. Break-resistant acrylic body with silk-screened scale. Unaffected by dilute acids and bases (except hydrofluoric acid and ammonium hydroxide) with an operational range of 1–14 pH. Not for use with alcohol or organic solvents. Leakproof and lubrication-free Teflon® TFE stopcock plug.
Ordering information: Shipped individually packaged. Complete with acrylic body, polymethylpentene stopcock and tip, Teflon® TFE plug, three feet of NALGENE® 380More Product Information